Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum oxide, neutral, for TLC

For TLC

Manufacturer:  Alfa Aesar™ 4605636

Quantity 500g

 View more versions of this product

Catalog No. AA4605636


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1344-28-1
Al2O3
MFCD00003424
Aluminum oxide, Neutral
Al2O3
101.96
14,356

Specifications

1344-28-1
Al2O3
101.96
14,356
5 to 15 Micron APS
2980°C
2045°C
Aluminum oxide, Neutral
Powder
Al2O3
MFCD00003424
Odorless
1.765
White
500g

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit