Alfa Aesar™ Aluminum oxide, polymeric precursor, 10% w/v solution in toluene

Aluminum oxide sealants/films

Manufacturer: Alfa Aesar™ 42767AD

Quantity 50mL

 View more versions of this product

Catalog No. AA42767AD

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00003424
    UN Number UN1993
    Chemical Name or Material Aluminum oxide, Polymeric precursor

    Specifications

    Physical Form Semi-Solid/Viscous Liquid
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 50mL
    Concentration 10% w/v solution in toluene
    Sensitivity Moisture Sensitive
    UN Number UN1993
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    TSCA TSCA
    MDL Number MFCD00003424
    Solubility Information Soluble in hexane, toluene
    Chemical Name or Material Aluminum oxide, Polymeric precursor

    GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit