missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, polymeric precursor, 10% w/v solution in toluene

Aluminum oxide sealants/films

Manufacturer:  Alfa Aesar™ 42767AB

Quantity 10mL

 View more versions of this product

Catalog No. AA42767AB


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00003424
UN1993
Aluminum oxide, Polymeric precursor

Specifications

10% w/v solution in toluene
Moisture Sensitive
MFCD00003424
Soluble in hexane, toluene
10mL
Semi-Solid/Viscous Liquid
UN1993
Aluminum oxide, Polymeric precursor

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'