Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide, Powder, Chromatography, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]

High quality chemicals for laboratory and specialized industrial use

    Suitable for Chromatographic Use
Formula Weight 101.96
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 4.00kg
Quantity 2.5kg
Chemical Name or Material Aluminum Oxide
Grade BAKER ANALYZED™ Reagent