Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide, Powder, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1344-28-1
1344-28-1
Grade Reagent
CAS Max % 1
Iron (Fe) 0.0003
Loss on Ignition 0.05
Packaging Poly Bottle
Sulfate 5e-05
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.