Learn More
Alfa Aesar™ Aluminum oxide, Refractory Brushable Paint, Water-based
Manufacturer:  Alfa Aesar™ 44887A1
Quantity 1kg
 View more versions of this product
Catalog No. AA44887A1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
1344-28-1
MFCD00003424
UN1760
Aluminum oxide, Refractory Brushable Paint, Water-based
Specifications
1344-28-1
MFCD00003424
White
Aluminum oxide, Refractory Brushable Paint, Water-based
Paint
UN1760
1kg
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : TSCA