Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, single crystal, Alfa Aesar™

Click to view available options
Quantity:
1 pc.
5 pc.
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Physical Form 12.7mm dia x 3.2mm thick,random orientation
Packaging Shrink-wrapped
Quantity 5 pc.
Chemical Name or Material Aluminum oxide

EINECSNumber : 215-691-6

TSCA : Yes

Recommended Storage : Ambient temperatures