Alfa Aesar™ Aluminum oxide, single crystal

Manufacturer: Alfa Aesar™ 36649KG

Quantity 1 piece

 View more versions of this product

Catalog No. AA36649KG

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11092-32-3
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum oxide, Single crystal, 25.4mm dia x 0.5mm thick, random orientation

    Specifications

    MDL Number MFCD00003424
    CAS 11092-32-3
    Quantity 1 piece
    TSCA TSCA
    Chemical Name or Material Aluminum oxide, Single crystal, 25.4mm dia x 0.5mm thick, random orientation

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit