Learn More
Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, R Plane, Alfa Aesar™
Manufacturer:  Alfa Aesar 38500KT
 View more versions of this product
Catalog No. AA38500KT
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
Specifications
Specifications
Aluminum Oxide Substrate, R Plane
Al2O3
1 ea.
Polished Two Sides
MFCD00003424
Safety and Handling
Safety and Handling
EINECSNumber : 215-691-6
TSCA : Yes
Recommended Storage : Ambient temperatures