Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, M Plane, Thermo Scientific™

Specifications

Chemical Name or Material Aluminum Oxide Substrate, M Plane
Name Note Polished Two Sides
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA41430KT
View Documents
Thermo Scientific Chemicals
41430KT
1 Ea. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
AA41430KU
View Documents
Thermo Scientific Chemicals
41430KU
5 Ea. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aluminum Oxide Substrate, M Plane
Polished Two Sides
Al2O3
MFCD00003424
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit