Alfa Aesar™ Aluminum oxide substrate, 10x10x0.5mm, polished two sides, C plane

Manufacturer: Alfa Aesar™ 41428KU

Quantity 5 each

 View more versions of this product

Catalog No. AA41428KU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-28-1
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum oxide substrate, 10x10x0.5mm, polished two sides, C plane

    Specifications

    CAS 1344-28-1
    TSCA TSCA
    MDL Number MFCD00003424
    Quantity 5 each
    RTECS Number BD1200000
    Chemical Name or Material Aluminum oxide substrate, 10x10x0.5mm, polished two sides, C plane

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit