Alfa Aesar™ Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, C Plane, Alfa Aesar™

Manufacturer: Alfa Aesar™ 43248KU

Quantity 5 ea.

 View more versions of this product

Catalog No. AA43248KU

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Molecular Formula Al2O3
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum Oxide Substrate, C Plane, Polished One Side

    Specifications

    Quantity 5 ea.
    Molecular Formula Al2O3
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum Oxide Substrate, C Plane, Polished One Side

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit