Alfa Aesar™ Aluminum oxide, super activated, acidic, Grade I

Manufacturer: Alfa Aesar™ 4606530

Quantity 250g

 View more versions of this product

Catalog No. AA4606530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Linear Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide, Super activated, acidic
    Grade Grade I

    Specifications

    Formula Weight 101.96
    Particle Size 50 to 200 Micron APS
    Physical Form Powder
    Refractive Index 1.765
    Boiling Point 2980°C
    Melting Point 2045°C
    Molecular Formula Al2O3
    Linear Formula Al2O3
    MDL Number MFCD00003424
    CAS 1344-28-1
    Merck Index 14,356
    TSCA TSCA
    Odor Odorless
    Color White
    Quantity 250g
    Chemical Name or Material Aluminum oxide, Super activated, acidic
    Grade Grade I

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit