Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, White powder, 99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4465914

Quantity 25g

 View more versions of this product

Catalog No. AA4465914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1344-28-1
99% (Metals basis)
101.96
Aluminum oxide
Powder
Al2O3
MFCD00003424

Specifications

1344-28-1
99% (Metals basis)
101.96
Odorless
25g
Powder
Al2O3
MFCD00003424
White
Aluminum oxide
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit