Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium phosphate, pure, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
7784-30-7
AlO4P
121.95
ILRRQNADMUWWFW-UHFFFAOYSA-K
aluminum phosphate,aluminum monophosphate,phosphalugel,phosphaljel,monoaluminum phosphate,aluminum acid phosphate,aluphos,aluminophosphoric acid,aluminiumphosphat german,aluminum phosphate 1:1
64655
aluminum;phosphate
[O-]P(=O)([O-])[O-].[Al+3]
7784-30-7
AlO4P
121.95
ILRRQNADMUWWFW-UHFFFAOYSA-K
aluminum phosphate,aluminum monophosphate,phosphalugel,phosphaljel,monoaluminum phosphate,aluminum acid phosphate,aluphos,aluminophosphoric acid,aluminiumphosphat german,aluminum phosphate 1:1
64655
aluminum;phosphate
[O-]P(=O)([O-])[O-].[Al+3]
Additional Information Al on Al2O3: 38 to 43%, P on P2O5: 55 to 60%
Linear Formula AlPO4
Merck Index 15,352
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol,soluble in acids,alkali
Formula Weight 121.95
Physical Form Powder
Grade Pure
Packaging Glass Bottle
Solubility Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol, soluble in acids, alkali
Color White
Melting Point >1500.0°C
Quantity 500g
Chemical Name or Material Aluminium phosphate
Show More Show Less

EINECSNumber : 232-056-9