Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum phosphate, pure, Thermo Scientific Chemicals

Catalog Number p-4510636
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
7784-30-7
AlO4P
121.95
ILRRQNADMUWWFW-UHFFFAOYSA-K
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
64655
aluminum;phosphate
[O-]P(=O)([O-])[O-].[Al+3]
7784-30-7
AlO4P
121.95
ILRRQNADMUWWFW-UHFFFAOYSA-K
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
64655
aluminum;phosphate
[O-]P(=O)([O-])[O-].[Al+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >1500.0°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula AlPO4
Merck Index 15,352
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol, soluble in acids, alkali
Formula Weight 121.95
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Aluminum phosphate
Show More Show Less
EINECSNumber 232-056-9

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit