Alfa Aesar™ Aluminum plate, alloy 6061, 6.35mm (0.25 in.) thick

Manufacturer: Alfa Aesar™ 42126RF

Quantity 30x30cm

 View more versions of this product

Catalog No. AA42126RF

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00214039
    Chemical Name or Material Aluminum plate, Alloy 6061, 6.35mm (0.25 in.) thick

    Specifications

    Physical Form Plate
    MDL Number MFCD00214039
    Odor Odorless
    Weight ≈1542g/30x30cm
    TSCA TSCA
    Quantity 30x30cm
    Chemical Name or Material Aluminum plate, Alloy 6061, 6.35mm (0.25 in.) thick

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit