Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Potassium Sulfate, Crystal, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7784-24-9
7784-24-9
Grade ACS
CAS Max % 1
Ammonium 5e-05
Percent Purity 98 to 102%
Chloride 5 ppm
Heavy Metals 1e-05
Insoluble Matter 5e-05
Iron (Fe) 1e-05
Packaging Poly Pail
Sodium (Na) 0.0002
Quantity 12 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.