Aluminum Potassium Sulfate Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical

Packaging Poly Bottle
Quantity 3kg

 View more versions of this product

Catalog No. A601-3

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7784-24-9
    Physical Form Solid
    Assay Percent Range 98.0 to 102.0 %
    Molecular Formula AlKO8S2 · 12 H2O
    Linear Formula AlK(SO4)2·12H2O
    Formula Weight 474.39
    MDL Number MFCD00149143
    Synonym Potassium Alum
    Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate
    Assay 98.0 to 102.0%
    Grade Certified ACS Grade

    Specifications

    Formula Weight 474.39
    Melting Point 92°C
    Quantity 3kg
    Physical Form Solid
    Color White
    pH 3.0 to 3.5
    Molecular Formula AlKO8S2 · 12 H2O
    Heavy Metals (as Pb) 0.001% max.
    Identification Pass Test
    Sodium (Na) 0.02% max.
    Insoluble Matter 0.005% max.
    Iron (Fe) 0.001% max.
    Packaging Poly Bottle
    Ammonium 0.005% max.
    MDL Number MFCD00149143
    CAS 7784-24-9
    ChemAlert Storage Symbol Gray
    Synonym Potassium Alum
    Chloride 5ppm max.
    Assay Percent Range 98.0 to 102.0 %
    Linear Formula AlK(SO4)2·12H2O
    Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate
    Assay 98.0 to 102.0%
    Grade Certified ACS Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit