Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Powder, -40+325 mesh, 99.8% (Metals basis), Thermo Scientific™

Catalog Number AA0001036
Click to view available options
Quantity:
100 g
500 g
2 kg
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray powder
Assay Percent Range 99.8% (Metals basis)
Quantity 500 g
Ignition Point 400°C (752°F)
IUPAC Name aluminum
Mesh Size -40+325 Mesh
Odor Odorless
Sensitivity Water reactive
Form Powder
Chemical Name or Material Aluminum
Show More Show Less
Hazard Category H250-H261-H500
Hazard Statement Keep container tightly closed and dry.
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-072-3
RTECSNumber BD0330000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit