Aluminum Powder, spherical, 99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 10576A9

Quantity 10kg

 View more versions of this product

Catalog No. AA10576A9

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    Physical Form Spherical Powder
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum

    Specifications

    Physical Form Spherical Powder
    Quantity 10kg
    Color Silver-gray
    DOT Information DOT Class: 4.3, Packaging Group: II
    EINECS Number 231-072-3
    Ignition Point 400°C (752°F)
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00134029
    CAS 7429-90-5
    Odor Odorless
    Health Hazard 1 H250-H261; Catches fire spontaneously if exposed to air. In contact with water releases flammable gas.
    Appearance Silver Gray powder
    Chemical Name or Material Aluminum

    H250-H261; Catches fire spontaneously if exposed to air. In contact with water releases flammable gas.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit