Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 44173A7

Quantity 5kg

 View more versions of this product

Catalog No. AA44173A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    MDL Number MFCD00134029
    UN Number UN1396
    Chemical Name or Material Aluminum powder, spherical, APS 4.5 to 7μm

    Specifications

    Quantity 5kg
    Physical Form Powder
    Odor Odorless
    Color Gray
    TSCA TSCA
    CAS 7429-90-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H250-H261;
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gas.
    Sensitivity Moisture Sensitive
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ALUMINUM POWDER, UNCOATED
    RTECS Number BD0330000
    MDL Number MFCD00134029
    UN Number UN1396
    Health Hazard 3 GHS P Statement
    P210-P222-P231+P232-P370+P378b-P422a-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Aluminum powder, spherical, APS 4.5 to 7μm
    Assay 97.5%, (metals basis)

    GHS H Statement
    H250-H261;
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gas.

    GHS P Statement
    P210-P222-P231+P232-P370+P378b-P422a-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit