Alfa Aesar™ Aluminum rod, alloy 6061, 25.4mm (1.0 in.) dia.

Manufacturer: Alfa Aesar™ 42055BV

Quantity 30cm

 View more versions of this product

Catalog No. AA42055BV

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00214039
    Chemical Name or Material Aluminum rod, Alloy 6061, 25.4mm (1.0 in.) dia.

    Specifications

    Physical Form Rod
    MDL Number MFCD00214039
    Weight ≈13.7 g/cm
    TSCA TSCA
    Odor Odorless
    Quantity 30cm
    Chemical Name or Material Aluminum rod, Alloy 6061, 25.4mm (1.0 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit