Alfa Aesar™ Aluminum sec-butoxide, 95%

Manufacturer: Alfa Aesar™ 3072122

Quantity 100g

 View more versions of this product

Catalog No. AA3072122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2269-22-9
    Formula Weight 246.32
    MDL Number MFCD00009327
    UN Number UN1992
    Beilstein 3910908
    Synonym Aluminum tri-sec-butoxide
    Chemical Name or Material Aluminum sec-butoxide

    Specifications

    Formula Weight 246.32
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture Sensitive
    Boiling Point 200° to 206°C (30mmHg)
    MDL Number MFCD00009327
    Beilstein 3910908
    Synonym Aluminum tri-sec-butoxide
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Physical Form Liquid
    CAS 2269-22-9
    UN Number UN1992
    Solubility Information Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols
    Health Hazard 1 Warning
    Density 0.967g/mL
    TSCA TSCA
    Quantity 100g
    Refractive Index 1.4390
    Flash Point 27° (82°F)
    Health Hazard 2 GHS H Statement
    H226-H302-H312-H315-H319
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Odor Alcohol-like
    Chemical Name or Material Aluminum sec-butoxide
    Assay 95%

    GHS H Statement
    H226-H302-H312-H315-H319
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit