Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum sec-butoxide, 95%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 4371236

Quantity 500g

 View more versions of this product

Catalog No. AA4371236


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2269-22-9
246.32
UN1992
Aluminum tri-sec-butoxide
Al[OCH(CH3)CH2CH3]3
MFCD00009327
3910908
Aluminum sec-butoxide

Specifications

200° to 206°C (30mmHg)
500g
0.967g/mL
ChemSeal™ bottles
Moisture Sensitive
Al[OCH(CH3)CH2CH3]3
MFCD00009327
3910908
Aluminum sec-butoxide
2269-22-9
27°C (82°F)
Alcohol-like
1.4390
Liquid
246.32
UN1992
Aluminum tri-sec-butoxide
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit