Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum sec-butoxide, 95%

Manufacturer:  Alfa Aesar™ 3072109

Quantity 10g

 View more versions of this product

Catalog No. AA3072109


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2269-22-9
MFCD00009327
3910908
Aluminum sec-butoxide
246.32
UN1992
Aluminum tri-sec-butoxide

Specifications

200° to 206°C (30mmHg)
10g
0.967g/mL
1.4390
Liquid
MFCD00009327
3910908
Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols
95%
2269-22-9
27° (82°F)
Alcohol-like
Moisture Sensitive
246.32
UN1992
Aluminum tri-sec-butoxide
Aluminum sec-butoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit