Learn More
Brady™ Aluminum Security, Admittance & CCTV Sign: NO TRESPASSING ON THIS PROPERTY UNDER PENALTY OF THE LAW
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to security, admittance and CCTV systems.
Manufacturer:  Brady™ 95237
Catalog No. 19-112-721
Add to cart
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications
Specifications
Black/blue on white
1/Ea.
14 in. x 10 in. x 0.035 in.
10 in.
NO TRESPASSING ON THIS PROPERTY UNDER PENALTY OF THE LAW
0.035 in.
14 in.
Aluminum
Rectangle
Corner holes
230°F
-40°F
Non-adhesive