Alfa Aesar™ Aluminum selenide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 1313206

Quantity 5g

 View more versions of this product

Catalog No. AA1313206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1302-82-5
    Formula Weight 290.84
    MDL Number MFCD00014158
    UN Number UN3283
    Merck Index 14,361
    Chemical Name or Material Aluminum selenide

    Specifications

    Formula Weight 290.84
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Decomposes to Al203 and SeO2 in water
    Odor Odorless
    Sensitivity Moisture Sensitive
    UN Number UN3283
    TSCA TSCA
    Physical Form Pieces
    MDL Number MFCD00014158
    Quantity 5g
    CAS 1302-82-5
    Merck Index 14,361
    Health Hazard 1 Danger
    Particle Size 6.5mm & down
    Color Pale brown
    Density 3.437g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Aluminum selenide
    Assay 99%, (metals basis)

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit