Aluminum shot, 99.99% (metals basis), Alfa Aesar™

Spheroid shape pellet 3/8 in. ±1/4 in.

Manufacturer: Alfa Aesar 4500136

Quantity 500g

 View more versions of this product

Catalog No. AA4500136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    Physical Form Shot, Nominally 9.5mm (0.375 in.) Flattened/Spherical Shape
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula Al
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum

    Specifications

    Physical Form Shot, Nominally 9.5mm (0.375 in.) Flattened/Spherical Shape
    Quantity 500g
    Color Silver
    EINECS Number 231-072-3
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula Al
    MDL Number MFCD00134029
    CAS 7429-90-5
    Odor Odorless
    Appearance Solid in various forms
    Chemical Name or Material Aluminum

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit