Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum shot, 99.99% (metals basis), Puratronic™, Alfa Aesar™

Click to view available options
Quantity:
50g
250g
1kg
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
Quantity 250g
Density 2.7
Assay Percent Range 99.999% (metals basis)
Formula Weight 26.98
Physical Form Sheet
Chemical Name or Material Aluminum, shot

EINECSNumber : 231-072-3

RTECSNumber : BD0330000

TSCA : Yes

Recommended Storage : Ambient temperatures