Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Brady™ Aluminum Sign: PACEMAKER WEARER - DO NOT ENTER THIS ROOM. MICROWAVE OVENS IN USE.
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 144573
 View more versions of this product
Catalog No. 19-112-485
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Yellow, Red, Black on White
Rectangle
PACEMAKER WEARER - DO NOT ENTER THIS ROOM. MICROWAVE OVENS IN USE.
Aluminum
14 in. x 10 in. x 0.035 in.