Learn More
Brady™ Aluminum Sign: RESPIRATORS ARE REQUIRED AT ALL TIMES
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 140820
Catalog No. 19-111-606
Add to cart
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Blue on White
Rectangle
RESPIRATORS ARE REQUIRED AT ALL TIMES
Aluminum
23.875 in. x 6 in. x 0.035 in.