Learn More
Brady™ Aluminum Sign: HEARING PROTECTION MUST BE WORN AT ALL TIMES
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Supplier: Brady™ 140830
Safety Services Available
Overview
Logo/Customization
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Blue on White | |
HEARING PROTECTION MUST BE WORN AT ALL TIMES | |
Aluminum |
23.875 x 6 in. | |
Rectangular |