Alfa Aesar™ Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 3849222

Quantity 100g

 View more versions of this product

Catalog No. AA3849222

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00143595
    Chemical Name or Material Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length
    Grade Puratronic™

    Specifications

    Physical Form Slug
    TSCA TSCA
    MDL Number MFCD00143595
    Avg. Mol. Wt. or Mol. Wt. Range Al:Si; 96.5:3.5 approximate wt%
    Quantity 100g
    Chemical Name or Material Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit