Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%
Click to view available options
Quantity:
25g
100g
Specifications
Weight | 1.1g/cm3 to 1.7g/cm3 |
Quantity | 100g |
Concentration | Ag 19 to 21 wt% |
Mesh Size | −200 Mesh |
Odor | Odorless |
Physical Form | Powder, Density 1.1-1.7g/cm |
Percent Purity | 99.9% |
Chemical Name or Material | Aluminum, Silver coated powder |
Assay | (metals basis) |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures