Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum slug, 1.98mm (0.078 in.) dia. x 8.0mm (0.315 in.) length, 99.99% (metals basis)

Click to view available options
Quantity:
10g
50g
250g
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]

Evaporation or metallizing source material

Quantity 50g
Weight ≈140pc/10g
Odor Odorless
Physical Form ≈140pc/10g
Percent Purity 99.99%
Chemical Name or Material Aluminum slug, 1.98mm (0.078 in.) dia. x 8.0mm (0.315 in.) length
Assay (metals basis)

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures