Alfa Aesar™ Aluminum spheres, 7.94mm (0.31 in.) dia., 99% (metals basis)

Manufacturer: Alfa Aesar™ 42031KP

Quantity 500 pieces

 View more versions of this product

Catalog No. AA42031KP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    MDL Number MFCD00213972
    Chemical Name or Material Aluminum, 7.94mm (0.31 in.) dia.

    Specifications

    TSCA TSCA
    Physical Form Spheres
    MDL Number MFCD00213972
    CAS 7429-90-5
    Avg. Mol. Wt. or Mol. Wt. Range Alloy 1100, Al:Cu; ≥99:0.12 wt%
    Quantity 500 pieces
    Chemical Name or Material Aluminum, 7.94mm (0.31 in.) dia.
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit