Learn More
Aluminum Standard for AAS, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 06154100ML
 View more versions of this product
Catalog No. 11-102-0648
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
7446-70-0
MFCD00003422
Aluminum chloride solution
Analytical Standard
Analytical Standard
AlCl3
1.0 g/L Al in hydrochloric acid
133.34
100 mL
Safety and Handling
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label