Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Standard for AAS, MilliporeSigma™ Supelco™

Specifications

CAS 7446-70-0
Grade Analytical Standard
Linear Formula AlCl3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-102-0648
View Documents
MilliporeSigma Supelco
06154100ML
100 mL
Each for $22.05
N/A
Due to product restrictions, please to purchase or view availability for this product.
11-101-9921
View Documents
MilliporeSigma Supelco
06154500ML
500 mL
Each for $59.80
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

7446-70-0
Analytical Standard
AlCl3
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit