Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Stearate, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
637-12-7
C54H105AlO6
877.41
CEGOLXSVJUTHNZ-UHFFFAOYSA-K
aluminium(3+) trioctadecanoate
[Al+3].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
637-12-7
C54H105AlO6
877.41
CEGOLXSVJUTHNZ-UHFFFAOYSA-K
aluminium(3+) trioctadecanoate
[Al+3].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

A1113, 637-12-7, C54H105O6Al

Spectrum™ Aluminum Stearate is also known as aluminum monostearate and is a salt of aluminum and stearic acid. It is used as a viscosity-increasing agent in non-aqueous cosmetic and pharmaceutical formulations. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 100%
Packaging Fiber Drum
Quantity 45 kg