Alfa Aesar™ Aluminum sulfate, anhydrous, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4456322

Quantity 100g

 View more versions of this product

Catalog No. AA4456322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10043-01-3
    Molecular Formula Al2(SO4)3
    Formula Weight 342.15
    MDL Number MFCD00003423
    Merck Index 14,366
    Chemical Name or Material Aluminum sulfate, Anhydrous

    Specifications

    Formula Weight 342.15
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003423
    Molecular Formula Al2(SO4)3
    CAS 10043-01-3
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    RTECS Number BD1700000
    TSCA TSCA
    Physical Form Powder
    Melting Point 770°C
    Density 2.71g/mL
    Sensitivity Hygroscopic
    Merck Index 14,366
    pH 3.5
    Storage Note 1
    • Packaged under Argon
    Chemical Name or Material Aluminum sulfate, Anhydrous
    Assay 99.99%, (metals basis)

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit