Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Sulfate, Crystal, FCC, 99.5-114%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7784-31-8
Al2H18O21S3
504.27
KFCWRPFNNIEZRY-UHFFFAOYSA-H
dialuminium(3+) nonahydrate trisulfate
O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
7784-31-8
Al2H18O21S3
504.27
KFCWRPFNNIEZRY-UHFFFAOYSA-H
dialuminium(3+) nonahydrate trisulfate
O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

A1114, 7784-31-8, Al2(SO4)3•(14-18)H2O

Spectrum™ Aluminum Sulfate, Crystal, FCC is soluble in water and is mainly used as a thickener and emulsifier in food preparation. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN3077
Percent Purity 99.5 to 114%
Fluoride 0.003%
Lead (Pb) 3 mg/kg
Packaging Poly Pail
Selenium (Se) 0.003%
Quantity 2.5 kg