Aluminum sulfate hydrate, 97+%, Alfa Aesar

Manufacturer: Alfa Aesar 12362A7

Quantity 5kg

 View more versions of this product

Catalog No. AA12362A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17927-65-0
    Physical Form Crystalline
    Assay Percent Range >97%
    Molecular Formula Al2(SO4)3·xH2O
    Formula Weight 342.15
    MDL Number MFCD00149136
    Synonym Alum
    Chemical Name or Material Aluminum sulfate hydrate, hydrate

    Specifications

    Formula Weight 342.15
    Quantity 5kg
    Color White
    EINECS Number 233-135-0
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range >97%
    Molecular Formula Al2(SO4)3·xH2O
    Appearance White crystalline
    MDL Number MFCD00149136
    CAS 17927-65-0
    Synonym Alum
    Health Hazard 1 H318-H315-H335
    Chemical Name or Material Aluminum sulfate hydrate, hydrate

    H318-H315-H335

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit