Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum sulfate hydrate, ACS, 98.0-102%

Click to view available options
Quantity:
250g
1kg
17927-65-0
Al2H2O13S3
360.146
MFCD00149136
BUACSMWVFUNQET-UHFFFAOYSA-H
aluminium sulfate hydrate,aluminum sulfate hydrate,dialuminum trisulfate hydrate,aluminum sulfate usp,aluminum sulfate 2:3 hydrate,alminum sulfate hydrate,aluminum sulfate usan,sulfuric acid, aluminum salt 3:2 , hydrate,2al.3so4.h2o
3080712
dialuminum;trisulfate;hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
17927-65-0
Al2H2O13S3
360.146
MFCD00149136
BUACSMWVFUNQET-UHFFFAOYSA-H
aluminium sulfate hydrate,aluminum sulfate hydrate,dialuminum trisulfate hydrate,aluminum sulfate usp,aluminum sulfate 2:3 hydrate,alminum sulfate hydrate,aluminum sulfate usan,sulfuric acid, aluminum salt 3:2 , hydrate,2al.3so4.h2o
3080712
dialuminum;trisulfate;hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Merck Index 14,366
Solubility Information It is slightly soluble in alcohol and dilute mineral acids.
Formula Weight 342.15 (Anhydrous)
Physical Form Crystalline
Percent Purity 98.0 to 102%
Grade ACS
Density 1.61 g/mL
Odor Odorless
pH 3.5
Quantity 1kg
Chemical Name or Material Aluminum sulfate hydrate
Show More Show Less

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P280-P305+P351+P338-P310

H318

EINECSNumber : 233-135-0

RTECSNumber : BD1700000

TSCA : Yes

Recommended Storage : Ambient temperatures