Alfa Aesar™ Aluminum sulfate hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1062906

Quantity 5g

 View more versions of this product

Catalog No. AA1062906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17927-65-0
    Formula Weight 342.15 (Anhydrous)
    MDL Number MFCD00149136
    Merck Index 14,366
    Chemical Name or Material Aluminum sulfate hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 342.15 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Crystalline
    CAS 17927-65-0
    Color White
    pH 3.5
    Quantity 5g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    Density 1.61g/mL
    MDL Number MFCD00149136
    Merck Index 14,366
    RTECS Number BD1700000
    Chemical Name or Material Aluminum sulfate hydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit