Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Sulfate, USP, 54-59%, Spectrum™

Click to view available options
Quantity:
100 lbs.
12 kg
2.5 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
17927-65-0
Al2H2O13S3
360.15
BUACSMWVFUNQET-UHFFFAOYSA-H
dialuminium(3+) hydrate trisulfate
O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
17927-65-0
Al2H2O13S3
360.15
BUACSMWVFUNQET-UHFFFAOYSA-H
dialuminium(3+) hydrate trisulfate
O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

AL300, 17927-65-0, Al2(SO4)3•xH2O

Spectrum™ Aluminum Sulfate, USP is a water soluble inorganic chemical compound. Uses in the pharmaceutical area are generally because of its astringent and hemostatic properties. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 54 to 59%
Packaging Poly Bottle
pH Range 2.9
Quantity 500 g