Alfa Aesar™ Aluminum trifluoromethanesulfonate

Manufacturer: Alfa Aesar™ 3856818

Quantity 50g

 View more versions of this product

Catalog No. AA3856818

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 74974-61-1
    Linear Formula Al(CF3SO3)3
    Formula Weight 474.2
    MDL Number MFCD00143596
    UN Number UN3261
    Synonym Aluminum triflate
    Chemical Name or Material Aluminum trifluoromethanesulfonate

    Specifications

    Formula Weight 474.2
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Physical Form Powder
    MDL Number MFCD00143596
    CAS 74974-61-1
    Synonym Aluminum triflate
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 50g
    Health Hazard 1 Danger
    Color White
    Sensitivity Hygroscopic
    Linear Formula Al(CF3SO3)3
    UN Number UN3261
    Melting Point ≈300°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Aluminum trifluoromethanesulfonate

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit