Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum Ultrathin foil, 0.0075 micron thick, 99.995% (metals basis)

Microelectronics, contacts and coatings

Manufacturer:  Alfa Aesar™ 40682DJ

Quantity 50mm

 View more versions of this product

Catalog No. AA40682DJ


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7429-90-5
MFCD00134029
Aluminum Ultrathin foil

Specifications

50mm
Circular Disc on Permanent Mylar Support
Odorless
Aluminum Ultrathin foil
7429-90-5
MFCD00134029
0.0075 Micron thick
99.995%, (metals basis)

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit