Alfa Aesar™ Aluminum Ultrathin foil, 1.0 micron thick, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 40684DH

Quantity 25mm

 View more versions of this product

Catalog No. AA40684DH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum Ultrathin foil

    Specifications

    Physical Form Circular Disc on Permanent Mylar Support
    Particle Size 1.0 Micron thick
    TSCA TSCA
    MDL Number MFCD00134029
    CAS 7429-90-5
    Odor Odorless
    Quantity 25mm
    Color Silver
    RTECS Number BD0330000
    Chemical Name or Material Aluminum Ultrathin foil
    Assay 99.995%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit