Learn More
Brady™ Aluminum Warning Sign: KEEP OUT HAZARD AREA
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$24.70 - $43.60
Specifications
Shape | Rectangular |
---|---|
Displayed Text | KEEP OUT HAZARD AREA |
Temperature Range (English) | -40°F to 230°F |
Finish | Not Laminated |
Material | Aluminum |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-107-001
|
Brady™
40677 |
10 x 7 in. |
Each for $24.70
|
|
|||||
19-107-002
|
Brady™
40678 |
14 x 10 in. |
Each for $43.60
|
|
|||||
Description
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor and outdoor use
Specifications
Rectangular | |
-40°F to 230°F | |
Aluminum | |
Non-adhesive | |
230°F | |
0.035 in. |
KEEP OUT HAZARD AREA | |
Not Laminated | |
1 Ea. | |
Corner holes | |
-40°F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.